3,4-Dihydro-2H-1,4-benzothiazine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3,4-Dihydro-2H-benzo[b][1,4]thiazine
H61045-03
H61045-06
AAH61045-03EA
114.08
CAD
AAH61045-03
AAH61045-06
3,4-Dihydro-2H-1,4-benzothiazine 97%
3,4-Dihydro-2H-1,4-benzothiazine
Purity: 97%
CAS No.: 3080-99-7
Molecular Formula: C8H9NS
Formula Weight: 151.23
MDL No.: MFCD04038593
Formula:
C₈H₉NS MW: 151.227 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04038593 CAS Number: 3080-99-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...