3-(Diphenylphosphino)propylamine ≥97%

Supplier: Thermo Scientific Chemicals
Danger

H60672-03 H60672-MD
AAH60672-03EA 218.04 CAD
AAH60672-03 AAH60672-06 AAH60672-MD
3-(Diphenylphosphino)propylamine ≥97%
3-(Diphenylphosphino)propylamine

Purity: 97+% CAS No.: 16605-03-1 Molecular Formula: C15H18NP Formula Weight: 243.28 Beilstein Reference: 2730850 UN No.: 2735 MDL No.: MFCD00233832

Formula: C₁₅H₁₈NP
Storage Temperature: Ambient
MDL Number: MFCD00233832
CAS Number: 16605-03-1
UN: 2735
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR