n-Dodecyl acrylate 90% stabilized, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning


Stabilization: Stabilised with 4-Methoxyphenol (MEHQ)

H60625-AE H60625-AP
AAH60625-AEEA 73.25 CAD
AAH60625-AE AAH60625-AP
n-Dodecyl acrylate 90% stabilized, Technical Grade
n-Dodecyl acrylate

Stabilised with with 60-100ppm 4-methoxyphenol


Purity: 90% CAS No.: 2156-97-0 Molecular Formula: C15H28O2 Formula Weight: 240.38 Flash Point: 110°(230°F) Refractive Index: 1.4450 Boiling Point: 120°/0.75mm Beilstein Reference: 1779069 UN No.: 3082 MDL No.: MFCD00042871

Formula: C₁₅H₂₈O₂
Boiling Pt: 120 °C/0.75 mm
Density: 0.884
Flash Pt: 110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042871
CAS Number: 2156-97-0
EINECS: 218-463-4
UN: 3082
ADR: "9",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR