2-(Diisopropylphosphino)ethylamine 10% (w/w) in THF

Supplier: Thermo Scientific Chemicals
Danger

H60435-06 H60435-14
AAH60435-06EA 135.63 CAD
AAH60435-06 AAH60435-14
2-(Diisopropylphosphino)ethylamine 10% (w/w) in THF
2-(Diisopropylphosphino)ethylamine

CAS No.: 1053657-14-9 Molecular Formula: C8H20NP Formula Weight: 161.22 UN No.: 2924 MDL No.: MFCD11044863

Formula: C₈H₂₀NP
MW: 161.229 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11044863
CAS Number: 1053657-14-9
UN: 2924
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR