Dimethylthiocarbamoyl chloride 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Dimethylthiocarbamoylchloride
H60104-14
H60104-22
AAH60104-14EA
124.98
CAD
AAH60104-14
AAH60104-22
Dimethylthiocarbamoyl chloride 95%
Dimethylthiocarbamoyl chloride
Purity: 95%
CAS No.: 16420-13-6
Molecular Formula: C3H6ClNS
Formula Weight: 123.60
Storage Temperature: 2°C to 8°C
Flash Point: 98°(208°F)
Melting Point: 39-43°
Boiling Point: 90-95°
Beilstein Reference: 635823
UN No.: 3261
MDL No.: MFCD00004919
Formula:
C₃H₆ClNS Boiling Pt: 90…95 °C Melting Pt: 39…43 °C Flash Pt: 98 °C(208 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004919 CAS Number: 16420-13-6 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...