4-Methyl-2-oxovaleric acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-Methyl-2-oxopentanoic Acid, 2-Keto-4-methylpentanoic Acid
H60076-03
H60076-06
AAH60076-03EA
108.7
CAD
AAH60076-03
AAH60076-06
4-Methyl-2-oxovaleric acid 98%
4-Methyl-2-oxovaleric acid
Purity: 98%
CAS No.: 816-66-0
Molecular Formula: C6H10O3
Formula Weight: 130.14
Storage Temperature: 2°C to 8°C
Refractive Index: 1.431
Melting Point: 8-10°
Boiling Point: 82-83°
Beilstein Reference: 1701823
UN No.: 3265
MDL No.: MFCD00066204
Formula:
C₆H₁₀O₃ MW: 130.14 g/mol Boiling Pt: 82…83 °C Melting Pt: 8…10 °C Density: 1.055 Storage Temperature: Ambient |
MDL Number:
MFCD00066204 CAS Number: 816-66-0 EINECS: 212-435-5 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...