2-Bromo-1-methylbenzimidazole 97%

Supplier: Thermo Scientific Chemicals
Warning

H60075-03 H60075-06 H60075-MD
AAH60075-03EA 290.72 CAD
AAH60075-03 AAH60075-06 AAH60075-MD
2-Bromo-1-methylbenzimidazole 97%
2-Bromo-1-methylbenzimidazole

Purity: 97% CAS No.: 49572-60-3 Molecular Formula: C8H7BrN2 Formula Weight: 211.06 MDL No.: MFCD00839528

Formula: C₈H₇BrN₂
MW: 211.062 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00839528
CAS Number: 49572-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR