2-Bromo-4-chlorobenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H59777-03 H59777-06
AAH59777-03EA 62.57 CAD
AAH59777-03 AAH59777-06
2-Bromo-4-chlorobenzoic acid 97%
2-Bromo-4-chlorobenzoic acid

Purity: 97% CAS No.: 936-08-3 Molecular Formula: C7H4BrClO2 Formula Weight: 235.46 Physical Form: Solid Melting Point: 157-161° UN No.: 2811 MDL No.: MFCD00672930

Formula: C₇H₄BrClO₂
Melting Pt: 157…161 °C
Storage Temperature: Ambient
MDL Number: MFCD00672930
CAS Number: 936-08-3
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR