Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N1-tert-butoxycarbonyl-L-tryptophan 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H59663-06
H59663-14
AAH59663-06EA
161.95
CAD
AAH59663-06
AAH59663-14
Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N1-tert-butoxycarbonyl-L-tryptophan 97%
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N1-tert-butoxycarbonyl-L-tryptophan
Purity: 97%
CAS No.: 143824-78-6
Molecular Formula: C31H30N2O6
Formula Weight: 526.58
Storage Temperature: 2°C to 8°C
Melting Point: ca 97° dec.
Beilstein Reference: 7698009
Optical Rotation: -21° (c=1 in DMF)
UN No.: 3077
MDL No.: MFCD00153366
Formula:
C₃₁H₃₀N₂O₆ Melting Pt: ca 97 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00153366 CAS Number: 143824-78-6 UN: 3077 ADR: "9",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...