4,4′-Azo-bis(4-cyanovaleric acid), (∼18% H₂O) 98%
Supplier: Thermo Scientific Chemicals
4,4'-Azobis(4-cyanovaleric acid) is used as a free radical imitator in polymer synthesis such as polyvinyl chloride, polyacrylonitrile, polyvinyl alcohol, and synthetic fibres. It is utilized for both heterogeneous and homogenous free-radical polymerizations and as an initiator in reversible addition-fragmentation chain transfer polymerization (RAFT). It is also used as a blowing agent for plastics and elastomers.
- Free radical imitator
- Blowing agent for plastics and elastomers
Purity: 98%
CAS No.: 2638-94-0
Molecular Formula: C12H16N4O4
Formula Weight: 280.28
Storage Temperature: 2°C to 8°C
Melting Point: 118-125° dec.
Beilstein Reference: 1729856
UN No.: 1325
MDL No.: MFCD00002799
Caution: Light senstive. Incompatible with strong oxidizing agents.
Formula:
C₁₂H₁₆N₄O₄ Melting Pt: 118…125 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00002799 CAS Number: 2638-94-0 EINECS: 220-135-0 ADR: 4.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...