3,4-Dichloro-6-fluoroaniline 96%

Supplier: Thermo Scientific Chemicals
Warning

H59506-03 H59506-MD H59506-03 H59506-MD
CAAAH59506-03EA 377.62 CAD
CAAAH59506-03 CAAAH59506-MD AAH59506-03 AAH59506-MD
3,4-Dichloro-6-fluoroaniline 96%
3,4-Dichloro-6-fluoroaniline

Purity: 96% CAS No.: 2729-36-4 Molecular Formula: C6H4Cl2FN Formula Weight: 180.01 UN No.: 2811 MDL No.: MFCD19707630

Formula: C₆H₄Cl₂FN
Storage Temperature: Ambient
MDL Number: MFCD19707630
CAS Number: 2729-36-4
UN: 2811
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR