6-bromoquinoline-5-carbonitrile 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H59484-03
H59484-MD
AAH59484-03EA
229.1
CAD
AAH59484-03
AAH59484-MD
6-bromoquinoline-5-carbonitrile 95%
6-bromoquinoline-5-carbonitrile
Purity: 95%
CAS No.: 1188365-70-9
Molecular Formula: C10H5BrN2
Formula Weight: 233.07
Beilstein Reference: 20114547
UN No.: 3439
MDL No.: MFCD22683072
Formula:
C₁₀H₅BrN₂ MW: 233.068 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22683072 CAS Number: 1188365-70-9 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...