1-(4-Bromo-3-fluorophenyl)cyclopropanecarbonitrile 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H59323-03
H59323-MD
AAH59323-03EA
636.74
CAD
AAH59323-03
AAH59323-MD
1-(4-Bromo-3-fluorophenyl)cyclopropanecarbonitrile 96%
1-(4-Bromo-3-fluorophenyl)cyclopropanecarbonitrile
Purity: 96%
CAS No.: 749269-73-6
Molecular Formula: C10H7BrFN
Formula Weight: 240.07
UN No.: 3439
MDL No.: MFCD19692057
Formula:
C₁₀H₇BrFN Storage Temperature: Ambient |
MDL Number:
MFCD19692057 CAS Number: 749269-73-6 UN: 3439 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...