1-Adamantanethiol 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Adamantane-1-thiol, 1-Mercaptoadamantane
H58958-03
H58958-MD
AAH58958-03EA
182.62
CAD
AAH58958-03
AAH58958-MD
1-Adamantanethiol 98%
1-Adamantanethiol
Purity: 98%
CAS No.: 34301-54-7
Molecular Formula: C10H16S
Formula Weight: 168.30
Melting Point: 99-106°
UN No.: 2811
MDL No.: MFCD00213530
Formula:
C₁₀H₁₆S Melting Pt: 99…106 °C Storage Temperature: Ambient |
MDL Number:
MFCD00213530 CAS Number: 34301-54-7 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...