3-(Methylamino)-1-propanol 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H58818-03
H58818-06
AAH58818-03EA
98.43
CAD
AAH58818-03
AAH58818-06
3-(Methylamino)-1-propanol 95%
3-(Methylamino)-1-propanol
Purity: 95%
CAS No.: 42055-15-2
Molecular Formula: C4H11NO
Formula Weight: 89.14
Storage Temperature: 2°C to 8°C
Flash Point: 82°(180°F)
Refractive Index: 1.4400
Boiling Point: 169°
UN No.: 2735
MDL No.: MFCD06797456
Formula:
C₄H₁₁NO Boiling Pt: 169 °C Density: 0.98 Flash Pt: 82 °C(180 °F) Storage Temperature: Ambient |
MDL Number:
MFCD06797456 CAS Number: 42055-15-2 UN: 2735 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...