(R)-(+)-Butylene oxide 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H58579-03
H58579-06
AAH58579-03EA
85.79
CAD
AAH58579-03
AAH58579-06
(R)-(+)-Butylene oxide 98%
(R)-(+)-Butylene oxide
Purity: 98%
CAS No.: 3760-95-0
Molecular Formula: C4H8O
Formula Weight: 72.11
Flash Point: -12°(10°F)
Refractive Index: 1.3810
Boiling Point: 60-64°
Optical Rotation: +10° (neat)
UN No.: 3022
MDL No.: MFCD02683444
Formula:
C₄H₈O Boiling Pt: 60…64 °C Density: 0.83 Flash Pt: -12 °C(10 °F) Storage Temperature: Ambient |
MDL Number:
MFCD02683444 CAS Number: 3760-95-0 UN: 3022 ADR: "3",II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...