(R)-(-)-2-Methylpyrrolidine 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(R)-2-Methylpyrrolidine
H57885-03
H57885-MD
AAH57885-03EA
230.68
CAD
AAH57885-03
AAH57885-MD
(R)-(-)-2-Methylpyrrolidine 95%
(R)-(-)-2-Methylpyrrolidine
Purity: 95%
CAS No.: 41720-98-3
Molecular Formula: C5H11N
Formula Weight: 85.15
Storage Temperature: 2°C to 8°C
Flash Point: 7°(45°F)
Refractive Index: 1.4300
Optical Rotation: -19° (c=5 in methanol)
UN No.: 2733
MDL No.: MFCD07783026
Formula:
C₅H₁₁N Density: 0.84 Flash Pt: 7 °C(45 °F) Storage Temperature: Ambient |
MDL Number:
MFCD07783026 CAS Number: 41720-98-3 UN: 2733 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...