(R)-(+)-2-Aminomethyl-1-ethylpyrrolidine 95%

Supplier: Thermo Scientific Chemicals
Danger

H57638-03 H57638-MD
AAH57638-03EA 151.68 CAD
AAH57638-03 AAH57638-MD
(R)-(+)-2-Aminomethyl-1-ethylpyrrolidine 95%

Purity: 95% CAS No.: 22795-97-7 Molecular Formula: C7H16N2 Formula Weight: 128.22 Flash Point: 57°(135°F) Refractive Index: 1.4670 Boiling Point: 50-54° Optical Rotation: +95° (c=1 in methanol) UN No.: 1993 MDL No.: MFCD00671482

Formula: C₇H₁₆N₂
Boiling Pt: 50…54 °C
Density: 0.929
Flash Pt: 57 °C(135 °F)
Storage Temperature: Ambient
MDL Number: MFCD00671482
CAS Number: 22795-97-7
UN: 2734
ADR: "8",II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR