3-bromooxetane 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Bromooxetane

H57437-MD H57437-ME
AAH57437-MDEA 162.74 CAD
AAH57437-MD AAH57437-ME
3-bromooxetane 95%
3-bromooxetane

Purity: 95% CAS No.: 39267-79-3 Molecular Formula: C3H5BrO Formula Weight: 136.98 Storage Temperature: 2°C to 8°C Flash Point: 48°(118°F) Boiling Point: 131° UN No.: 1993 MDL No.: MFCD08544402

Formula: C₃H₅BrO
MW: 136.976 g/mol
Boiling Pt: 131 °C
Density: 1.793
Flash Pt: 48 °C (118 °F)
Storage Temperature: Ambient
MDL Number: MFCD08544402
CAS Number: 39267-79-3
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR