3-(Methylamino)oxetane 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H57425-MD
H57425-ME
AAH57425-MDEA
534.04
CAD
AAH57425-MD
AAH57425-ME
3-(Methylamino)oxetane 95%
3-(Methylamino)oxetane
Purity: 95%
CAS No.: 952182-03-5
Molecular Formula: C4H9NO
Formula Weight: 87.12
Storage Temperature: 2°C to 8°C
Flash Point: 40°(104°F)
Physical Form: Liquid
Appearance: Colorless to pale yellow
UN No.: 1993
MDL No.: MFCD08544349
Formula:
C₄H₉NO Flash Pt: 40 °C(104 °F) Storage Temperature: Ambient |
MDL Number:
MFCD08544349 CAS Number: 952182-03-5 UN: 1993 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...