(S)-(+)-1-Boc-3-bromopyrrolidine 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H57408-03
H57408-MD
AAH57408-03EA
260.7
CAD
AAH57408-03
AAH57408-MD
(S)-(+)-1-Boc-3-bromopyrrolidine 95%
(S)-(+)-1-Boc-3-bromopyrrolidine
Purity: 95%
CAS No.: 569660-89-5
Molecular Formula: C9H16BrNO2
Formula Weight: 250.14
Storage Temperature: 2°C to 8°C
UN No.: 3077
MDL No.: MFCD17214728
Formula:
C₉H₁₆BrNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD17214728 CAS Number: 569660-89-5 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...