5-Amino-2-(trifluoromethyl)pyridine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Amino-6-(trifluoromethyl)pyridine, 2-(Trifluoromethyl)pyridin-5-amine, 2-(Trifluoromethyl)-5-pyridylamine

H56968-03 H56968-06
AAH56968-03EA 114.87 CAD
AAH56968-03 AAH56968-06 AAH56968-MD
5-Amino-2-(trifluoromethyl)pyridine 98%
5-Amino-2-(trifluoromethyl)pyridine

Purity: 98% CAS No.: 106877-33-2 Molecular Formula: C6H5F3N2 Formula Weight: 162.11 Flash Point: 110°(230°F) Melting Point: 39-43° UN No.: 2811 MDL No.: MFCD01862656

Formula: C₆H₅F₃N₂
MW: 162.11 g/mol
Melting Pt: 39…43 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD01862656
CAS Number: 106877-33-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR