tert-Butyl-3-butenoate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H56859-03
H56859-06
AAH56859-03EA
170.64
CAD
AAH56859-03
AAH56859-06
tert-Butyl-3-butenoate 97%
tert-Butyl-3-butenoate
Purity: 97%
CAS No.: 14036-55-6
Molecular Formula: C8H14O2
Formula Weight: 142.20
Flash Point: 31°(88°F)
Refractive Index: 1.4130
UN No.: 3272
MDL No.: MFCD06411714
Formula:
C₈H₁₄O₂ Density: 0.877 Flash Pt: 31 °C(88 °F) Storage Temperature: Ambient |
MDL Number:
MFCD06411714 CAS Number: 14036-55-6 UN: 3272 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...