4-Chloro-7-azaindole 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H56734-03
H56734-MD
AAH56734-03EA
153.58
CAD
AAH56734-03
AAH56734-06
AAH56734-MD
4-Chloro-7-azaindole 98%
4-Chloro-7-azaindole
Purity: 98%
CAS No.: 55052-28-3
Molecular Formula: C7H5ClN2
Formula Weight: 152.58
Melting Point: 176-181°
UN No.: 2811
MDL No.: MFCD08272232
Formula:
C₇H₅ClN₂ Melting Pt: 176…181 °C Storage Temperature: Ambient |
MDL Number:
MFCD08272232 CAS Number: 55052-28-3 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...