Cyclopentanecarbaldehyde 98% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Stabilization:
Stabilised with 1,4-benzochinon 25 ppm
H56609-03
H56609-06
AAH56609-03EA
67.78
CAD
AAH56609-03
AAH56609-06
Cyclopentanecarbaldehyde 98% stabilized
Cyclopentanecarbaldehyde
Purity: 98%
CAS No.: 872-53-7
Molecular Formula: C5H9CHO
Formula Weight: 98.14
Flash Point: 28°(82°F)
Refractive Index: 1.4430
Boiling Point: 140-141°
UN No.: 1989
MDL No.: MFCD00060798
Formula:
C₆H₁₀O Boiling Pt: 140…141 °C Density: 0.919 Flash Pt: 28 °C(82 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00060798 CAS Number: 872-53-7 EINECS: 212-829-7 UN: 1989 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...