Undecanoyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Undecanoyl Chloride, Undecanoylchloride
H56380-03
H56380-ME
AAH56380-03EA
279.66
CAD
AAH56380-03
AAH56380-ME
Undecanoyl chloride 98%
Undecanoyl chloride
Purity: 98%
CAS No.: 17746-05-3
Molecular Formula: C11H21ClO
Formula Weight: 204.74
Storage Temperature: -30°C to -10°C
Flash Point: 113°(235°F)
Refractive Index: 1.4430
Boiling Point: 135-136°
Beilstein Reference: 1759226
UN No.: 3265
MDL No.: MFCD00000739
Formula:
C₁₁H₂₁ClO Boiling Pt: 135…136 °C Flash Pt: 113 °C (235 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000739 CAS Number: 17746-05-3 EINECS: 241-741-1 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...