1-Boc-4-(2-hydroxyethyl)piperazine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: tert-Butyl 4-(2-hydroxyethyl)piperazine-1-carboxylate, 4-(2-Hydroxyethyl)piperazine, N1-BOC protected

H55991-06 H55991-14
AAH55991-03EA 40.92 CAD
AAH55991-03 AAH55991-06 AAH55991-14
1-Boc-4-(2-hydroxyethyl)piperazine 97%
1-Boc-4-(2-hydroxyethyl)piperazine

Purity: 97% CAS No.: 77279-24-4 Molecular Formula: C11H22N2O3 Formula Weight: 230.31 Flash Point: 110°(230°F) Physical Form: Solid Melting Point: 38-42° UN No.: 2811 MDL No.: MFCD00728947

Formula: C₁₁H₂₂N₂O₃
Melting Pt: 38…42 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00728947
CAS Number: 77279-24-4
UN: 3259
ADR: 8,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR