4-Iodo-2-(trifluoromethyl)benzonitrile 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H55609-03
H55609-06
AAH55609-03EA
66.83
CAD
AAH55609-03
AAH55609-06
AAH55609-MD
4-Iodo-2-(trifluoromethyl)benzonitrile 97%
4-Iodo-2-(trifluoromethyl)benzonitrile
Purity: 97%
CAS No.: 101066-87-9
Molecular Formula: C8H3F3IN
Formula Weight: 297.02
Flash Point:
Formula:
C₈H₃F₃IN Melting Pt: 50…55 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD08458091 CAS Number: 101066-87-9 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...