Tri-tert-butyl borate 97%

Supplier: Thermo Scientific Chemicals
Warning

H55461-06 H55461-14
AAH55461-06EA 109.87 CAD
AAH55461-06 AAH55461-14
Tri-tert-butyl borate 97%
Tri-tert-butyl borate

Purity: 98% CAS No.: 7397-43-5 Molecular Formula: C12H27BO3 Formula Weight: 230.16 Flash Point: 29°(84°F) Physical Form: Liquid Density: 0.811 Refractive Index: 1.3890 Appearance: Colorless Melting Point: 18-19° Boiling Point: 101°/74mm UN No.: 1993 MDL No.: MFCD00009652

Formula: C₁₂H₂₇BO₃
Boiling Pt: 101 °C/74 mm
Melting Pt: 18…19 °C
Density: 0.811
Flash Pt: 29 °C(84 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009652
CAS Number: 7397-43-5
UN: 1993
ADR: "3",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR