Cryptocyanine 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H55456-03
H55456-06
H55456-MD
AAH55456-03EA
191.18
CAD
AAH55456-03
AAH55456-06
AAH55456-MD
Cryptocyanine 96%
Cryptocyanine
Purity: 96%
CAS No.: 4727-50-8
Molecular Formula: C25H25IN2
Formula Weight: 480.38
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: Green
Melting Point: 250-253°
UN No.: 2811
MDL No.: MFCD00011970
Formula:
C₂₅H₂₅IN₂ Melting Pt: 250…253 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011970 CAS Number: 4727-50-8 EINECS: 225-224-8 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...