1-(4-Chlorobenzyl)piperazine 98%

Supplier: Thermo Scientific Chemicals
Danger

H55337-03 H55337-06 H55337-14
AAH55337-03EA 55.77 CAD
AAH55337-03 AAH55337-06 AAH55337-14
1-(4-Chlorobenzyl)piperazine 98%
1-(4-Chlorobenzyl)piperazine

Purity: 98% CAS No.: 23145-88-2 Molecular Formula: C11H15ClN2 Formula Weight: 210.71 Flash Point: 110°(230°F) Physical Form: Liquid Density: 1.140 Refractive Index: 1.5580 Appearance: Colorless to pale yellow Boiling Point: 104-106°/0.1mm UN No.: 3267 MDL No.: MFCD00040791

Formula: C₁₁H₁₅ClN₂
Boiling Pt: 104…106 °C/0.1 mm
Density: 1.140
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040791
CAS Number: 23145-88-2
UN: 2735
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR