2-(Tributylstannyl)furan 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H55259-AA
H55259-AC
AAH55259-AAEA
64.78
CAD
AAH55259-AA
AAH55259-AC
AAH55259-AE
2-(Tributylstannyl)furan 97%
2-(Tributylstannyl)furan
Purity: 97%
CAS No.: 118486-94-5
Molecular Formula: C16H30OSn
Formula Weight: 357.10
Physical Form: Liquid
Density: 1.134
Refractive Index: 1.4940
Appearance: Colorless to yellow
Boiling Point: 90°/0.1mm
UN No.: 2788
MDL No.: MFCD00192512
Formula:
C₁₆H₃₀OSn Boiling Pt: 90 °C/0.1 mm Density: 1.134 Storage Temperature: Ambient |
MDL Number:
MFCD00192512 CAS Number: 118486-94-5 UN: 2788 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...