4-Dimethylaminopyridinium bromide perbromide 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-(Dimethylamino)pyridine tribromide

H55010-06 H55010-14
AAH55010-06EA 72.05 CAD
AAH55010-06 AAH55010-14 AAH55010-22
4-Dimethylaminopyridinium bromide perbromide 97%
4-Dimethylaminopyridinium bromide perbromide

Purity: 97% CAS No.: 92976-81-3 Molecular Formula: C7H10N2.HBr.Br2 Formula Weight: 362.89 Melting Point: 129-132° UN No.: 3261 MDL No.: MFCD00043439

Formula: C₇H₁₁Br₃N₂
MW: 362.89 g/mol
Melting Pt: 129…132 °C
Storage Temperature: Ambient
MDL Number: MFCD00043439
CAS Number: 92976-81-3
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR