2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H54410-03
H54410-06
AAH54410-03EA
225.94
CAD
AAH54410-03
AAH54410-06
AAH54410-MD
2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride 97%
2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-sulfonyl chloride
Purity: 97%
CAS No.: 28485-18-9
Molecular Formula: C4H3ClN2O4S
Formula Weight: 209.95
UN No.: 3261
MDL No.: MFCD06655485
Formula:
C₄H₃ClN₂O₄S Storage Temperature: Ambient |
MDL Number:
MFCD06655485 CAS Number: 28485-18-9 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...