Piperidine-1-sulfonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

H54053-03 H54053-06 H54053-MD
AAH54053-03EA 205.4 CAD
AAH54053-03 AAH54053-06 AAH54053-MD
Piperidine-1-sulfonyl chloride 97%
Piperidine-1-sulfonyl chloride

Purity: 97% CAS No.: 35856-62-3 Molecular Formula: C5H10ClNO2S Formula Weight: 183.65 Refractive Index: 1.4920

Formula: C₅H₁₀ClNO₂S
Density: 1.308
Storage Temperature: Ambient
MDL Number: MFCD03250329
CAS Number: 35856-62-3
UN: 3265
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR