Praseodymium trichloride, ultra dry 99.95% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
H37815-06
H37815-14
AAH37815-03EA
106.4
CAD
AAH37815-03
AAH37815-06
AAH37815-14
Praseodymium trichloride, ultra dry 99.95% (REO, rare earth oxide basis)
Praseodymium trichloride
Purity: 99.95%
CAS No.: 10361-79-2
Molecular Formula: Cl3Pr
Formula Weight: 247.27
Physical Form: Powder, ampouled under argon
Density: 4.02
Melting Point: 786°
Boiling Point: 1700°
MDL No.: MFCD00011176
Formula:
Cl₃Pr Boiling Pt: 1700 °C Melting Pt: 786 °C Density: 4.02 Storage Temperature: Ambient |
MDL Number:
MFCD00011176 CAS Number: 10361-79-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...