3-Bromo-1H-indazole 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H37775-03
H37775-09
AAH37775-03EA
99.38
CAD
AAH37775-03
AAH37775-09
3-Bromo-1H-indazole 97%
3-Bromo-1H-indazole
Purity: 97%
CAS No.: 40598-94-5
Molecular Formula: C7H5BrN2
Formula Weight: 197.04
Melting Point: 148-149°
UN No.: 2811
MDL No.: MFCD00159926
Formula:
C₇H₅BrN₂ Melting Pt: 148…149 °C Storage Temperature: Ambient |
MDL Number:
MFCD00159926 CAS Number: 40598-94-5 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...