5-Methylthiophene-2-carbonyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

H36784-03 H36784-MD
AAH36784-03EA 368.14 CAD
AAH36784-03 AAH36784-MD
5-Methylthiophene-2-carbonyl chloride 98%
5-Methylthiophene-2-carbonyl chloride

Purity: 98% CAS No.: 31555-59-6 Molecular Formula: C6H5ClOS Formula Weight: 160.62 Physical Form: Liquid Sensitivity: Moisture Sensitive Appearance: Colorless to pale yellow Boiling Point: 78-80°/0.75mm UN No.: 3265

Formula: C₆H₅ClOS
MW: 160.615 g/mol
Boiling Pt: 78…80 °C/0.75 mm
Storage Temperature: Ambient
MDL Number: MFCD00130092
CAS Number: 31555-59-6
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR