3,4-Dihydroquinolin-2(1H)-one 98%

Supplier: Thermo Scientific Chemicals
Warning

H36638-03 H36638-06
AAH36638-03EA 73.79 CAD
AAH36638-03 AAH36638-06 AAH36638-14
3,4-Dihydroquinolin-2(1H)-one 98%
3,4-Dihydroquinolin-2(1H)-one

Purity: 98% CAS No.: 553-03-7 Molecular Formula: C9H9NO Formula Weight: 147.18 Melting Point: 165-167° MDL No.: MFCD00016722

Formula: C9H9NO
Melting Pt: 165…167 °C
Storage Temperature: Ambient
MDL Number: MFCD00016722
CAS Number: 553-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR