1,3-Dicyclohexylimidazolium tetrafluoroborate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H36633-03
H36633-06
AAH36633-03EA
141.25
CAD
AAH36633-03
AAH36633-06
1,3-Dicyclohexylimidazolium tetrafluoroborate 97%
1,3-Dicyclohexylimidazolium tetrafluoroborate
Purity: 97%
CAS No.: 286014-38-8
Molecular Formula: C15H25BF4N2
Formula Weight: 320.18
Physical Form: Powder
Appearance: White to pale brown
Melting Point: 173-177°C
MDL No.: MFCD04973310
Formula:
C₁₅H₂₅BF₄N₂ Melting Pt: 173…177°C Storage Temperature: Ambient |
MDL Number:
MFCD04973310 CAS Number: 286014-38-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...