3,5-Bis(methoxycarbonyl)benzeneboronic acid pinacol ester 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H36525-03
H36525-06
AAH36525-03EA
282.82
CAD
AAH36525-03
AAH36525-06
AAH36525-MD
3,5-Bis(methoxycarbonyl)benzeneboronic acid pinacol ester 97%
3,5-Bis(methoxycarbonyl)benzeneboronic acid pinacol ester
Purity: 97%
CAS No.: 944392-68-1
Molecular Formula: C16H21BO6
Formula Weight: 320.15
Physical Form: Solid
Appearance: Pale pink
Melting Point: 128°
MDL No.: MFCD11858596
Formula:
C₁₆H₂₁BO₆ Melting Pt: 128 °C Storage Temperature: Ambient |
MDL Number:
MFCD11858596 CAS Number: 944392-68-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...