Methyl-5-methylpyrazole-3-carboxylate 96%

Supplier: Thermo Scientific Chemicals

Synonyms: Methyl 5-methyl-1H-pyrazole-3-carboxylate, Methyl 5-methyl[1H]pyrazole-3-carboxylate, 5-Methylpyrazole-3-carboxylic Acid Methyl Ester

H36130-03 H36130-06
AAH36130-03EA 92.11 CAD
AAH36130-03 AAH36130-06
Methyl-5-methylpyrazole-3-carboxylate 96%
Methyl-5-methylpyrazole-3-carboxylate

Purity: 96% CAS No.: 25016-17-5 Molecular Formula: C6H8N2O2 Formula Weight: 140.14 Melting Point: 79-81° MDL No.: MFCD03778987

Formula: C₆H₈N₂O₂
MW: 140.14 g/mol
Melting Pt: 79…81 °C
Storage Temperature: Ambient
MDL Number: MFCD03778987
CAS Number: 25016-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR