2-Chloro-5-nitrobenzophenone 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
(2-Chloro-5-nitrophenyl)(phenyl)methanone
H35800-14
H35800-22
AAH35800-14EA
102.54
CAD
AAH35800-14
AAH35800-22
2-Chloro-5-nitrobenzophenone 98%
2-Chloro-5-nitrobenzophenone
Purity: 98%
CAS No.: 34052-37-4
Molecular Formula: C13H8ClNO3
Formula Weight: 261.66
Melting Point: 78-84°
MDL No.: MFCD00007295
Formula:
C₁₃H₈ClNO₃ MW: 261.66 g/mol Melting Pt: 78…84 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007295 CAS Number: 34052-37-4 EINECS: 251-811-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...