3-(Aminomethyl)phenol 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Hydroxybenzylamine

H35708-03 H35708-06 H35708-MD
AAH35708-MDEA 88.05 CAD
AAH35708-MD AAH35708-06 AAH35708-03
3-(Aminomethyl)phenol 97%
3-(Aminomethyl)phenol
Formula: C₇H₉NO
Melting Pt: 170…173 °C
Storage Temperature: Ambient
MDL Number: MFCD00798977
CAS Number: 73604-31-6
UN: 3259
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR