1H-Pyrrole-3-carbaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Pyrrole-3-carboxaldehyde
H35559-03
H35559-MD
AAH35559-03EA
425.02
CAD
AAH35559-03
AAH35559-MD
1H-Pyrrole-3-carbaldehyde 97%
1H-Pyrrole-3-carbaldehyde
Purity: 97%
CAS No.: 7126-39-8
Molecular Formula: C5H5NO
Formula Weight: 95.10
Melting Point: 65-68°
MDL No.: MFCD03426733
Formula:
C5H5NO MW: 95.10 g/mol Melting Pt: 65…68 °C Storage Temperature: Ambient |
MDL Number:
MFCD03426733 CAS Number: 7126-39-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...