2-Chloro-6-nitrobenzothiazole 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Chloro-6-nitrobenzo[d]thiazole, 2-Chloro-6-nitro-1,3-benzothiazole
H35525-03
H35525-09
AAH35525-03EA
51.82
CAD
AAH35525-03
AAH35525-09
2-Chloro-6-nitrobenzothiazole 95%
2-Chloro-6-nitrobenzothiazole
Purity: 95%
CAS No.: 2407-11-6
Molecular Formula: C7H3ClN2O2S
Formula Weight: 214.63
Melting Point: 185-189°
MDL No.: MFCD00022852
Formula:
C₇H₃ClN₂O₂S Melting Pt: 185…189 °C Storage Temperature: Ambient |
MDL Number:
MFCD00022852 CAS Number: 2407-11-6 EINECS: 219-302-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...