Ethyl potassium oxalate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H35403-06
H35403-14
AAH35403-06EA
170.64
CAD
AAH35403-06
AAH35403-14
Ethyl potassium oxalate 97%
Ethyl potassium oxalate
Purity: 97%
CAS No.: 1906-57-6
Molecular Formula: C4H5O4K
Formula Weight: 156.18
Melting Point: 220-223°
UN No.: 2811
MDL No.: MFCD00045861
Formula:
C₄H₅KO₄ Melting Pt: 220…223 °C Storage Temperature: Ambient |
MDL Number:
MFCD00045861 CAS Number: 1906-57-6 EINECS: 217-606-8 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...