6-Fluoro-1,2,3,4-tetrahydroquinoline 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H35364-03
H35364-MD
AAH35364-03EA
443.98
CAD
AAH35364-03
AAH35364-MD
6-Fluoro-1,2,3,4-tetrahydroquinoline 97%
6-Fluoro-1,2,3,4-tetrahydroquinoline
Purity: 97%
CAS No.: 59611-52-8
Molecular Formula: C9H10FN
Formula Weight: 151.18
Melting Point: 32-34°
MDL No.: MFCD09040997
Formula:
C₉H₁₀FN Melting Pt: 32…34 °C Storage Temperature: Ambient |
MDL Number:
MFCD09040997 CAS Number: 59611-52-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...