1-Methyl-3-octylimidazolium bis(trifluoromethylsulfonyl)imide 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 1-Methyl-3-n-octylimidazolium Bis(trifluoromethanesulfonyl)imide, 1-Methyl-3-n-octylimidazolium Bis(trifluoromethanesulphonyl)imide

H35352-06 H35352-14 H35352-22
AAH35352-06EA 129.05 CAD
AAH35352-06 AAH35352-14 AAH35352-22
1-Methyl-3-octylimidazolium bis(trifluoromethylsulfonyl)imide 99%
1-Methyl-3-octylimidazolium bis(trifluoromethylsulphonyl)imide

Purity: 99% CAS No.: 178631-04-4 Molecular Formula: C14H23F6N3O4S2 Formula Weight: 475.46 MDL No.: MFCD08458946

Formula: C₁₄H₂₃F₆N₃O₄S₂
MW: 475.48 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08458946
CAS Number: 178631-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR