2,4-Dimethyl-1,3-pentadiene 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H35300-06
H35300-14
AAH35300-06EA
138.25
CAD
AAH35300-06
AAH35300-14
2,4-Dimethyl-1,3-pentadiene 98%
2,4-Dimethyl-1,3-pentadiene
Purity: 98%
CAS No.: 1000-86-8
Molecular Formula: C7H12
Formula Weight: 96.17
Flash Point: 10°(50°F)
Refractive Index: 1.4400
Boiling Point: 94-96°
UN No.: 3295
MDL No.: MFCD00008903
Formula:
C₇H₁₂ Boiling Pt: 94…96 °C Density: 0.744 Flash Pt: 10 °C (50 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008903 CAS Number: 1000-86-8 EINECS: 213-677-4 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...